Learn More
Medchemexpress LLC Becotatug (Synonyms: JMT-101) 5mg
SDP
Supplier:  Medchemexpress LLC HYP9900495MG
Encompass_Preferred
Becotatug (JMT-101) is an IgG1 antibody targeting EGFR that can also be conjugated to Afatinib (HY-10261) and Osimertinib (HY-15772) as a synthetic ADC.
Catalog No. NC3397880