Learn More
Medchemexpress LLC HY-14881 10mg Medchemexpress, Bedaquiline CAS:843663-66-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1488110MG
Encompass_Preferred
Medchemexpress, HY-14881 10mg Bedaquiline CAS:843663-66-1 Bedaquiline (TMC207) is a diarylquinoline drug and inhibits Mycobacterium tuberculosis (Mtb) F1FO-ATP synthase through targeting of both the c- and the ε-subunit. Bedaquiline has uncoupler activity. Bedaquiline is used for the multi-drug resistant tuberculosis[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9074