Learn More
Medchemexpress LLC Bedaquiline fumarate | 845533-86-0 | 99.99% | 671.58 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14881A100MG
Encompass_Preferred
Bedaquiline fumarate is a diarylquinoline antibiotic that targets ATP synthase and is effective for treating *Mycobacterium tuberculosis* infections. It inhibits the growth of TDR *M. tuberculosis* strains with MIC values ranging from 0.125 to 0.5 mg/L, and also demonstrates high activity against *Mycobacterium avium* and moderate activity against other nontuberculous mycobacteria (NTM) species.
  • Inhibits TDR *M. tuberculosis* strains with MIC values from 0.125 to 0.5 mg/L
  • Highest activity against *Mycobacterium avium* among slowly growing mycobacteria (SGM)
  • More susceptible against *Mycobacterium abscessus* subsp. *abscessus* and *massiliense* than *Mycobacterium fortuitum* among rapidly growing mycobacteria (RGM)
  • Shows moderate in vitro activity against NTM species
  • Excellent in vitro activity against multidrug-resistant *Mycobacterium tuberculosis*
Catalog No. 50-163-8219