Learn More
Medchemexpress LLC Bedaquinoline impurity 1 | 1972612-60-4 | 98.6% | 555.50 | 100 MG

Supplier: Medchemexpress LLC HY14881E100MG
Bedaquinoline impurity 1 is an impurity of Bedaquinoline.
- Purity: 98.63%
- Molecular weight: 555.50
- Formula: C32H31BrN2O2
- Appearance: Solid
- Storage: Powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month
- For research use only
- Not sold to patients
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.