Learn More
Medchemexpress LLC Befiradol hydrochloride | 2436760-81-3 | 99.9% | 430.32 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14785A50MG
Encompass_Preferred
Befiradol hydrochloride (NLX-112 hydrochloride) is a selective 5-HT1A receptor agonist. It is intended for research use only.
  • Selective 5-HT1A receptor agonist
  • Intended for research use only
Catalog No. 50-003-61597