Learn More
Waters Corp ACQUITY UPLC Protein BEH SEC Column, 200Å, 1.7 µm, 4.6 mm X 150 mm, 10K - 500K, 1/pk
Supplier:  Waters Corp 186005225
Encompass_Preferred
Based upon the highly successful BEH particle technology, the new patent pending ACQUITY UPLC BEH200 SEC column represents the first commer­cially available sub 2 ìm SEC chemistry. Fully optimized for the analysis of proteins and their aggregates with molecular weights ranging from 10,000 to 450,000 Daltons, the total system solution will routinely deliver fast, accurate aggregate determinations up to 10 times quicker that traditional HPLC-based SEC methods.
Catalog No. NC9020419