Learn More
Selleck Chemical LLC Behenic Acid S5381-25mg
SDP
Supplier:  Selleck Chemical LLC S538125MG
Encompass_Preferred
Behenic Acid (docosanoic acid) is a long-chain saturated fatty acid that exists naturally as a triglyceride in canola It act as a lubricant that helps restore the skin s natural oils and improve overall levels of hydration
Catalog No. 50-311-0328