Learn More
Medchemexpress LLC Belantamab, 10mg, CAS No. : 2061894-48-0
SDP
Supplier:  Medchemexpress LLC HYP998010MG
Encompass_Preferred
Belantamab (GSK2857914) is a humanised IgG1 anti-BCMA (TNFRSF17) monoclonal antibody. Belantamab can be used in the synthesis of antibody-drug conjugate (ADC), Belantamab mafodotin.
Catalog No. NC3315730