Learn More
Medchemexpress LLC Belapectin 1mg | 1980787-47-0 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1144401MG
Encompass_Preferred
Belapectin is a carbohydrate-based galectin-3 inhibitor for preclinical research. It binds Gal-3 with a reported Ki of 2.8 μM, and is supplied as a solid with high purity.
  • Carbohydrate-based galectin-3 inhibitor
  • Reported binding affinity Ki = 2.8 μM
  • High purity (98.9%) suitable for research applications
  • Solid formulation with recommended cold storage
  • Available in small quantities for laboratory studies
Catalog No. 50-002-29991