Learn More
Apexbio Technology LLC Belinostat (PXD101), 50mg. Cas: 414864-00-9 MFCD: MFCD08064035. Hydroxamate-type HDAC inhibitor
SDP
Supplier:  Apexbio Technology LLC A409650
Encompass_Preferred
Belinostat (also known as PXD101) is a novel and potent hydroxamate-type inhibitor of histone deacetylase (HDAC) that inhibits the activity of HDAC in Hela cell extracts with 50% inhibition concentration IC50 value of 27 nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4878