Learn More
Cayman Chemical BelIzatInIb 25mg
SDP
Supplier:  Cayman Chemical 2845825 MG
Encompass_Preferred
An inhibitor of ALK and tropomyosin-related kinases A/B/C (IC50s = 0.7 and <3 nM, respectively)
Catalog No. 50-294-0337