Learn More
Selleck Chemical LLC Belizatinib (TSR-011) 5mg 1357920-84-3
SDP
Supplier:  Selleck Chemical LLC S85115MG
Encompass_Preferred
"Belizatinib (TSR-011) is a potent inhibitor of ALK (IC50=0.7 nM) and tropomyosin receptor kinase (TRK) (IC50 values less than 3 nM for TRK A, B, and C)." *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8372