Learn More
TARGETMOL CHEMICALS INC BELNACASAN 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T609025MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Belnacasan (VX-765) is an orally active IL-converting enzyme/caspase-1 inhibitor. purity: 99%
Catalog No. 50-257-9573