Learn More
Selleck Chemical LLC Belotecan
SDP
Supplier:  Selleck Chemical LLC S66315MG
Encompass_Preferred
Belotecan (CKD-602) is a potent DNA topoisomerase I inhibitor that exerts a clinical anticancer effect on various types of tumor
Catalog No. 50-000-00049