Learn More
TARGETMOL CHEMICALS INC BELOTECAN HYDROCHLORIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T755125MG
Encompass_Preferred
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Belotecan hydrochloride (CKD-602) is a synthetic water-soluble camptothecin derivative and topoisomerase I inhibitor with potential antitumor activitypurity: 98%
Catalog No. 50-257-9182