Learn More
Cayman Chemical BELVARAFENIB 10MG
SDP
Supplier:  Cayman Chemical 3146010 MG
Encompass_Preferred
An inhibitor of B-RAF and C-RAF (IC50s = 41, 7, and 2 nM for wild-type B-RAF, B-RAFV600E, and wild-type C-RAF, respectively); cytotoxic to B-RAFV600E-expressing A375 and SK-MEL-28 melanoma cells (IC50s = 57 and 69 nM, respectively), as well as COLO 205 and HCT116 colon and B-CPAP and CAL-62 thyroid cancer cells (IC50s = 116, 65, 43, and 479 nM, respectively); reduces tumor growth in A375, SK-MEL-28, SK-MEL-2, and SK-MEL-30 mouse xenograft models
Catalog No. 50-299-2100