Learn More
Selleck Chemical LLC Belvarafenib S8853-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S885310MM/1ML
Encompass_Preferred
Belvarafenib is a selective and orally bioavailable pan-RAF kinase inhibitor with IC50 values of 41 nM 7 nM and 2 nM for BRAF WT BRAF(V600E) and CRAF kinases respectively
Catalog No. 50-000-25421