Learn More
Selleck Chemical LLC Belzutifan PT2977-5mg
SDP
Supplier:  Selleck Chemical LLC S88865MG
Encompass_Preferred
Belzutifan (PT2977, MK-6482), an orally active and selective HIF-2α inhibitor, increases potency and improves pharmacokinetic profile, providing a potential treatment for clear cell renal cell carcinoma (ccRCC).
Catalog No. 50-238-9727