Learn More
Medchemexpress LLC Bemcentinib | 1037624-75-1 | MFCD21608463 | 99.9% | 506.6 g/mol | C30H34N8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15150R10MG
Encompass_Preferred
Bemcentinib analytical standard is a reference-grade Axl receptor tyrosine kinase inhibitor supplied for research and analytical applications. It is a selective, orally active compound with reported cellular IC50 ≈ 14 nM and demonstrated selectivity versus related kinases, suitable for assay development, compound characterization, and pharmacology studies.
  • High purity suitable for analytical use (reported HPLC purity 99.92%).
  • Selective Axl inhibitor with cellular IC50 ≈ 14 nM.
  • Supports assay development and compound characterization.
  • Solid analytical standard supplied in 10 MG packages.
  • Chemical formula C30H34N8, molecular weight 506.6 g/mol, CAS 1037624-75-1.
Catalog No. 50-002-86955