Learn More
Selleck Chemical LLC Bemcentinib (R428) S2841-1g
SDP
Supplier:  Selleck Chemical LLC S28411G
Encompass_Preferred
Bemcentinib (R428 BGB324) is an inhibitor of Axl with IC50 of 14 nM 100-fold selective for Axl versus Abl Selectivty for Axl is also greater than Mer and Tyro3 (50-to-100- fold more selective) and InsR EGFR HER2 and PDGFR (100- fold more selective)
Catalog No. 50-310-8170