Learn More
Apexbio Technology LLC Bemegride 64-65-3 50mg
SDP
Supplier:  Apexbio Technology LLC B167250
Encompass_Preferred
Bemegride (CAS 64-65-3) is a central nervous system stimulant that acts primarily as an antagonist of barbiturate-induced depression It functions by enhancing neuronal excitability counteracting the inhibitory effects of barbiturates on synaptic transmission Bemegride has been extensively utilized in experimental models to investigate CNS stimulation and mechanisms of drug-induced respiratory depression Its application as an antagonist in barbiturate toxicity studies makes it a valuable tool for evaluating neuropharmacological responses and exploring reversal strategies for sedative overdose in biomedical research
Catalog No. 50-224-0721