Learn More
Medchemexpress LLC HY-137958A 5mg , Bemnifosbuvir CAS:1998705-64-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY137958A5MG
Encompass_Preferred
HY-137958A 5mg Bemnifosbuvir CAS: 1998705-64-8 Bemnifosbuvir (AT-511) is a potent and orally active HCV viral replication inhibitor. Bemnifosbuvir is highly effective in the control of SARS-CoV-2 (COVID-19) infection in vitro (EC90=0.47 μM). Bemnifosbuvir has pangenotypic antiviral activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9720