Learn More
Selleck Chemical LLC Bemnifosbuvir Hemisulfate-25MG
SDP
Supplier:  Selleck Chemical LLC E101325MG
Encompass_Preferred
Bemnifosbuvir Hemisulfate (AT-527 RG-6422 RO 7496998) is a hemi-sulfate salt of AT-511 AT-511 is a potent inhibitor of SARS-CoV-2 with an EC90 of 0 47 M
Catalog No. 50-313-7166