Learn More
Medchemexpress LLC Bempedoic acid | 738606-46-7 | MFCD18800820 | 98.0% | 344.49 g·mol⁻¹ | C19H36O5 | 10MM 1ML

Supplier: Medchemexpress LLC HY1235710MM 1ML
Bempedoic acid (ETC-1002) is an ATP-citrate lyase (ACL) inhibitor used in biochemical and cellular research to modulate lipid metabolism and activate AMPK. The material is supplied as a powder and is commonly prepared as concentrated stocks in DMSO (for example, 10 mM). Intended for research use only.
- ATP-citrate lyase inhibitor and AMPK activator.
- Supplied as powder suitable for preparation of DMSO stock solutions.
- Molecular formula C19H36O5; molecular weight 344.49 g·mol⁻¹.
- Typical purity 98.0% (NMR) as reported on certificate of analysis.
- Recommended storage: powder -20 °C for long term; in solvent -80 °C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.