Learn More
Medchemexpress LLC Bempedoic acid-d4 | 2408131-70-2 | 98.0% | 348.51 g/mol | C19H32D4O5 | 1MG
SDP
Supplier:  Medchemexpress LLC HY12357S21MG
Encompass_Preferred
Bempedoic acid-d4 is the deuterium-labeled analog of bempedoic acid provided for research use as an analytical tracer and internal standard. It is intended for use in mass spectrometry quantitation and pharmacokinetic studies and corresponds to the biological target profile of bempedoic acid as an ATP-citrate lyase inhibitor with reported AMPK activity. Store and handle following the supplier datasheet recommendations.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for pharmacokinetic and tracer studies.
  • Retains ATP-citrate lyase inhibitory activity for biological reference.
  • High purity suitable for analytical applications.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-37660