Learn More
Medchemexpress LLC Bempedoic acid-d4 | 2408131-70-2 | 98.0% | 348.51 g/mol | C19H32D4O5 | 5MG

Supplier: Medchemexpress LLC HY12357S25MG
Bempedoic acid-d4 is the deuterium-labeled analogue of bempedoic acid supplied as a powder research standard for analytical and biochemical studies. It is used as an internal standard in quantitative assays and for metabolic pathway research related to ATP-citrate lyase and AMPK activity.
- deuterium-labeled internal standard for analytical assays
- molecular formula: C19H32D4O5
- molecular weight: 348.51 g/mol
- purity: 98.0%
- form: powder; solubility in DMSO 50 mg/mL (ultrasonic)
- storage: -20°C (3 years); 4°C (2 years) for powder
- typical package sizes: 1 mg, 5 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.