Learn More
Apexbio Technology LLC Benazepril 86541-75-5 25mg
SDP
Supplier:  Apexbio Technology LLC B127825
Encompass_Preferred
Benazepril (CAS 86541-75-5) is a small molecule inhibitor of angiotensin-converting enzyme (ACE) a key enzyme responsible for the conversion of angiotensin I to the vasoconstrictor angiotensin II By blocking ACE activity benazepril reduces the synthesis of angiotensin II leading to diminished vasoconstriction and lowered blood pressure In biomedical research benazepril is utilized to investigate the renin-angiotensin system cardiovascular regulation and mechanisms underlying hypertension Its pharmacological profile supports its application in studies focused on cardiovascular diseases and renal function
Catalog No. 50-000-88590