Learn More
Medchemexpress LLC Benazepril | 86541-75-5 | 99.85% | C24H28N2O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB009310MM/1ML
Encompass_Preferred
Benazepril is an orally active angiotensin-converting enzyme (ACE) inhibitor that reduces angiotensin-II production. It inhibits oxidative stress and apoptosis via the PI3K/Akt signaling pathway. This compound improves diabetic nephropathy, decreases proteinuria, and is used in the study of hypertension, heart failure, and diabetic nephropathy.
  • Orally active ACE inhibitor
  • Reduces angiotensin-II production
  • Inhibits oxidative stress and apoptosis via PI3K/Akt signaling pathway
  • Improves diabetic nephropathy
  • Decreases proteinuria
  • Used in the study of hypertension, heart failure, and diabetic nephropathy
  • Alleviates oxidative stress and inhibits cell apoptosis in H9c2 cells (in vitro)
  • Decreases proteinuria, urea, creatinine, triglycerides, and total cholesterol in diabetic nephropathy rat models (in vivo)
Catalog No. 50-004-32804