Learn More
Medchemexpress LLC Benazepril | 86541-75-5 | 99.9% | C24H28N2O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0093100MG
Encompass_Preferred
Benazepril is an orally active angiotensin-converting enzyme (ACE) inhibitor that effectively reduces angiotensin-II production. It inhibits oxidative stress and apoptosis through the PI3K/Akt signaling pathway, making it valuable in the study of hypertension, heart failure, and diabetic nephropathy. It has been shown to improve diabetic nephropathy and decrease proteinuria.
  • Orally active angiotensin-converting enzyme (ACE) inhibitor
  • Reduces angiotensin-II production
  • Inhibits oxidative stress and apoptosis via the PI3K/Akt signaling pathway
  • Improves diabetic nephropathy
  • Decreases proteinuria
  • Used in the study of hypertension, heart failure, and diabetic nephropathy
Catalog No. 50-004-32552