Learn More
Medchemexpress LLC Benazepril | 86541-75-5 | 99.9% | C24H28N2O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB009350MG
Encompass_Preferred
Benazepril is an orally active angiotensin-converting enzyme (ACE) inhibitor that reduces angiotensin-II production. It inhibits oxidative stress and apoptosis via the PI3K/Akt signaling pathway. Benazepril has been shown to improve diabetic nephropathy and decrease proteinuria.
  • Inhibits oxidative stress and apoptosis via the PI3K/Akt signaling pathway
  • Improves diabetic nephropathy
  • Decreases proteinuria
  • Used in the study of hypertension
  • Used in the study of heart failure
  • Used in the study of diabetic nephropathy
Catalog No. 50-004-32631