Learn More
Medchemexpress LLC Benazepril (CGS14824A free base) | 86541-75-5 | MFCD00864466 | 99.9% | 424.49 g/mol | C24H28N2O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB009310MG
Encompass_Preferred
Benazepril (CGS14824A free base) is an orally active angiotensin-converting enzyme (ACE) inhibitor used in research on hypertension, cardiovascular disease, and renal function. Supplied as a white to off-white solid and available in small solid packs or as a DMSO solution. Purity is listed at 99.85% and molecular weight is 424.49 g/mol.
  • Orally active ACE inhibitor used in hypertension and cardiovascular research.
  • Free base solid form; appearance white to off-white.
  • Molecular formula C24H28N2O5; molecular weight 424.49 g/mol.
  • High purity (99.85%) suitable for research applications.
  • Available as small solid packs (e.g., 10 mg) and solution format (10 mM in DMSO).
Catalog No. 50-002-99044