Learn More
Apexbio Technology LLC Benazepril HCl 86541-74-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B22165.1
Encompass_Preferred
Benazepril hydrochloride (CAS 86541-74-4) is a small molecule inhibitor targeting angiotensin converting enzyme (ACE) a key protease involved in converting angiotensin I to the bioactive vasoconstrictor angiotensin II By suppressing ACE activity benazepril HCl reduces angiotensin II-mediated vasoconstriction and aldosterone secretion leading to vascular relaxation and decreased blood pressure In biomedical research benazepril hydrochloride serves as a useful tool for investigating pathways involved in hypertension cardiovascular disorders and angiotensin signaling regulation
Catalog No. 50-000-61816