Learn More
Medchemexpress LLC Benazepril hydrochloride | 86541-74-4 | 99.96% | C24H29ClN2O5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0093A5G
Encompass_Preferred
Benazepril hydrochloride is an orally active angiotensin-converting enzyme (ACE) inhibitor that reduces angiotensin-II production. It inhibits oxidative stress and apoptosis via the PI3K/Akt signaling pathway. Additionally, Benazepril hydrochloride improves diabetic nephropathy and decreases proteinuria, making it suitable for studies of hypertension, heart failure, and diabetic nephropathy.
  • Orally active ACE inhibitor
  • Reduces angiotensin-II production
  • Inhibits oxidative stress and cell apoptosis
  • Improves diabetic nephropathy and decreases proteinuria
  • Used in studies of hypertension, heart failure, and diabetic nephropathy
Catalog No. 50-004-32879