Learn More
Medchemexpress LLC Benazepril hydrochloride | 86541-74-4 | CGS14824A | 99.96% | C24H29ClN2O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0093A10MM/1ML
Encompass_Preferred
Benazepril hydrochloride is an orally active angiotensin-converting enzyme (ACE) inhibitor that reduces angiotensin-II production. It inhibits oxidative stress and apoptosis via the PI3K/Akt signaling pathway. This compound can be used in studies of hypertension, heart failure, and diabetic nephropathy.
  • Targets angiotensin-converting enzyme (ACE), apoptosis, Akt, and reactive oxygen species
  • Reduces angiotensin-II production
  • Inhibits oxidative stress and apoptosis through PI3K/Akt signaling pathway
  • Improves diabetic nephropathy and decreases proteinuria
Catalog No. 50-004-32878