Learn More
Medchemexpress LLC Benazepril hydrochloride | 86541-74-4 | MFCD00895734 | 100.0% | C24H29ClN2O5 | 1 G
SDP
Supplier:  Medchemexpress LLC HYB0093A1G
Encompass_Preferred
Benazepril hydrochloride is an orally active angiotensin-converting enzyme (ACE) inhibitor that reduces angiotensin-II production. It inhibits oxidative stress and apoptosis via the PI3K/Akt signaling pathway and improves diabetic nephropathy and decreases proteinuria. It is used in the study of hypertension, heart failure, and diabetic nephropathy.
  • Orally active ACE inhibitor
  • Reduces angiotensin-II production
  • Inhibits oxidative stress and apoptosis
  • Improves diabetic nephropathy
  • Decreases proteinuria
  • White to off-white solid appearance
  • For research use only
Catalog No. 50-004-32632