Learn More
Grainger Bench Grinder 8 36 Grit 3/4HP
Supplier:  Grainger 16W006
Encompass_Preferred
The Dayton Bench Grinder Buffer is a fixed base buffer with a 6 inch diameter by 2 inch wide buffing wheel. It has a 1/3 HP, 115V AC, 1725 RPM motor and operates on standard household current. The buffer has an on/off toggle switch with rubber bumper guards for safety. It features a steel base with rubber feet to prevent walking or creeping during use. The Dayton buffer allows for buffing, polishing, and finishing of metal, plastic, and wood workpieces. This is a light duty buffer suitable for general purpose buffing applications.
Catalog No. 50-276-7743