Learn More
IW Tremont Co Inc LabExact Bench Protector Paper, spill containment, 48cm x 100 meter Roll
SDP
Supplier:  IW Tremont Co Inc LEBPR4650M
Encompass_Preferred
LabExact Disposable, highly absorbent pure cellulose paper with polyethylene backing is designed to protect laboratory surfaces against liquid spills and prevent leakage through to the bench top, necessary to maintain a safer work environment. Meets OSHA Regulation 29CFR 1910.1030 for Occupational Exposure to Bloodborne Pathogens. Tear resistant material, whether wet or dry. The polyethylene backing material is skid resistant, waterproof and chemical resistant. The absorbent paper quickly traps spilled or drained fluids. Smooth white surface is easy to write on. Roll, 48cm x 50m
Catalog No. 50-223-7869