Learn More
Grainger BenchPedestal Rctngl Slvr 16 1/4 x15
Supplier:  Grainger 14L053
Encompass_Preferred
The Grainger Approved Bench Pedestal Grinder is a 1/3 HP, 115V, 1725 RPM grinder with a 6 inch wheel diameter and 1 inch arbor size. It has a cast iron base with rubber feet to reduce vibration and movement. The motor is totally enclosed fan cooled for durability. It comes with eye shields, spark deflectors, and tool rests. This grinder is ideal for sharpening tools and deburring metal.
Catalog No. 50-275-3116