Learn More
Sigma Aldrich Fine Chemicals Biosciences BenchRockerTM variable 2D1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7554861EA
Encompass
The BenchRocker 2D is designed to provide the precise speeds and tilt angles required for a broad range of molecular and biological mixing applications. The user can adjust both parameters to match the vessel size and the volume of liquid being mixed yielding optimum results.
Catalog No. 50-290-8551