Learn More
JDM LAB SOLUTIONS LLC BENCHTOP INCUBATOR SHAKER
Supplier:  JDM LAB SOLUTIONS LLC ISRSDAA
Encompass
Benchtop Incubator Shaker IS-RSDAAFeatures Versatile platform with a wide selection of accessories Precise temperature control (ambient +5C~ 80C Speeds of up to 300 RPM with an orbit diameter of 26mm (1.02?) The Sliding platform feature provides easy sample handling with Max Load 2x1L Fits conveniently under countertopsTemperature Range: 5 C - 80CSpeed range: 0 - 300 RPMPlatform Dimensions: 13.8 x 11.8 inchesWeight: 110 pounds
Catalog No. NC2307320