Learn More
Medchemexpress LLC HY-B2141 500mg Medchemexpress, Bendazol CAS:621-72-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2141500MG
Encompass_Preferred
Medchemexpress, HY-B2141 500mg Bendazol CAS:621-72-7 Bendazol is a hypotensive drug which can also enhance NO synthase activity in renal glomeruli and collecting tubules. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2726