Learn More
Medchemexpress LLC Benfluorex hydrochloride | 23642-66-2 | 99.85% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1058500MG
Encompass_Preferred
Benfluorex hydrochloride (JP-992 hydrochloride) is a hepatic nuclear factor 4 alpha (HNF4α) activator. It is for research use only.
  • Consistently activates insulin promoter activity, increasing GFP-positive cells and endogenous insulin mRNA levels.
  • Stimulates HNF4α expression and alters HNF4α protease sensitivity.
  • Decreases the synthesis of acid-soluble products and ketone bodies from oleate.
  • Markedly increases the production of 14CO2 into the citric acid cycle.
  • Inhibits gluconeogenesis from lactate/pyruvate in a dose-dependent manner.
Catalog No. 50-004-11875