Learn More
Medchemexpress LLC S-Benzoylthiamine O-monophosphate | 22457-89-2 | 99.38% | 466.45 | 500MG
SDP
Supplier:  Medchemexpress LLC HY17374500MG
Encompass_Preferred
Benfotiamine is a vitamin B1 derivative that exhibits potent antioxidative and anti-inflammatory activity. It can be used for the research of various secondary diabetic complications. It also can be used in infectious diseases.
  • Exhibits potent antioxidative activity.
  • Exhibits anti-inflammatory activity.
  • Applicable for research into secondary diabetic complications.
  • Can be used in infectious disease research.
Catalog No. 50-003-84985