Learn More
Medchemexpress LLC Benorilate | 5003-48-5 | MFCD00864257 | 99.9% | 313.30 g/mol | C17H15NO5 | 25 G
SDP
Supplier:  Medchemexpress LLC HY10779525G
Encompass_Preferred
Benorilate is an esterification product of paracetamol and acetylsalicylic acid supplied for research and analytical applications. It exhibits anti-inflammatory, analgesic, and antipyretic activity and can inhibit prostaglandin synthesis.
  • High purity: 99.85%.
  • Molecular formula C17H15NO5 and molecular weight 313.30 g/mol.
  • Exhibits anti-inflammatory, analgesic, and antipyretic properties.
  • Intended for research and analytical applications.
  • Available in multiple pack sizes, including 25 G.
Catalog No. 50-002-42359