Learn More
TARGETMOL CHEMICALS INC BENPERIDOL 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T263605MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Benperidol (Anquil) is a butanone analog with antipsychotic and stabilizing properties and has been shown to be effective in blocking D2 receptors.Benperidol can be used in positron emission tomography (PET) scanning to measure D(2) dopamine receptor binding in vivo.Benperidol has been shown to be effective in blocking D2 receptors in vivo.Benperidol has been shown to be effective in blocking D2 receptors in vivo. purity: 97%
Catalog No. 50-258-0702