Learn More
Medchemexpress LLC HY-B0404A 500mg Medchemexpress, Benserazide (hydrochloride) CAS:14919-77-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0404A500MG
Encompass_Preferred
Medchemexpress, HY-B0404A 500mg Benserazide (hydrochloride) CAS:14919-77-8 Benserazide hydrochloride (Serazide) is commonly used in Parkinson's disease and is an inhibitor of peripheral aromatic L-amino acid decarboxylase (AADC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2522