Learn More
Medchemexpress LLC Benserazide hydrochloride | 14919-77-8 | 99.7% | 10 MM 1 ML
Supplier: Medchemexpress LLC HYB0404A10MM/1ML
Benserazide hydrochloride (Serazide) is an aromatic L-amino acid decarboxylase (AADC) and L-DOPA decarboxylase inhibitor. It also acts as a PKM2 inhibitor by directly binding to and blocking PKM2 enzyme activity, leading to inhibition of aerobic glycolysis and concurrent up-regulation of OXPHOS. This compound is suitable for research related to Parkinson's disease and melanoma.
- Acts as an aromatic L-amino acid decarboxylase (AADC) and L-DOPA decarboxylase inhibitor.
- Inhibits PKM2 enzyme activity.
- Inhibits aerobic glycolysis and up-regulates OXPHOS.
- Supports research in Parkinson's disease and melanoma.
- Inhibits protein aggregation and minimizes amyloid-induced cytotoxicity in human neuroblastoma cell line SH-SY5Y.
- Increases exogenous L-DOPA-derived extracellular DA levels in 6-OHDA-lesioned male Wistar rats.
- Reduces central AADC activity in the striatum of rats with nigrostriatal denervation.