Learn More
Medchemexpress LLC Benserazide (hydrochloride) | 14919-77-8 | MFCD00078571 | >98.0% | 5 G

Supplier: Medchemexpress LLC HYB0404A5G
Benserazide hydrochloride, also known as Serazide, is an aromatic L-amino acid decarboxylase (AADC) and L-DOPA decarboxylase inhibitor. It also acts as a PKM2 inhibitor by directly binding to and blocking PKM2 enzyme activity, which leads to the inhibition of aerobic glycolysis and concurrent up-regulation of OXPHOS. This compound is valuable in the study of Parkinson's disease and melanoma.
- Inhibits L-amino acid decarboxylase and L-DOPA decarboxylase.
- Acts as a PKM2 inhibitor by directly binding to and blocking its enzyme activity.
- Leads to inhibition of aerobic glycolysis and concurrent up-regulation of OXPHOS.
- Valuable in the study of Parkinson's disease.
- Applicable in melanoma studies.
- Target: Pyruvate kinase.
- Pathway: Metabolic enzyme/protease.
- Soluble in DMSO (100 mg/mL) and water (≥ 50 mg/mL).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.