Learn More
Cayman Chemical Bentamapimod 10MG
SDP
Supplier:  Cayman Chemical 2148210 MG
Encompass_Preferred
A selective, orally bioavailable inhibitor of JNK1, JNK2, and JNK3 (IC50s = 80, 90, and 230 nM, respectively); induces regression of endometriotic lesions without suppressing estrogen action in animal models of endometriosis
Catalog No. 50-298-6504