Learn More
TARGETMOL CHEMICALS INC Benthiazole 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T35735MG
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Benthiazole (Superdavloxan) is a broad-spectrum seed protectant that is effective against soil- and seed-borne fungal or bacterial diseases. Purity 87.65%
Catalog No. 50-287-8772